Key Documents
AXYRESV25S
Reservoir
capacity 25 mL, sterile
Synonym(s):
Liquid Reservoir, Reservoir Container, Storage Reservoir
About This Item
UNSPSC Code:
41121522
NACRES:
NB.13
Pricing and availability is not currently available.
sterility
sterile
manufacturer/tradename
Axygen Inc
Corning
capacity
25 mL
General description
  • Axygen® reagent reservoirs are optimized for use with multichannel pipettors.
  • The pour-off spouts on each corner and the sharp "V" bottom design facilitate maximum sample retrieval.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)