PHR3229
Proparacaine Hydrochloride
pharmaceutical secondary standard, certified reference material
Synonym(s):
Proparacaine hydrochloride, Proxymetacaine hydrochloride
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1571001
API family
proparacaine
CofA
current certificate can be downloaded
packaging
pkg of 500 mg
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
Cl.CCCOc1ccc(cc1N)C(=O)OCCN(CC)CC
InChI
1S/C16H26N2O3.ClH/c1-4-10-20-15-8-7-13(12-14(15)17)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H
InChI key
BFUUJUGQJUTPAF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service