Skip to Content
Merck
All Photos(1)

Key Documents

PHL89615

Cyanidin 3,5-diglucoside chloride

phyproof® Reference Substance

Synonym(s):

Cyanin chloride, Cyanidin-3,5-di-O-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClO16
CAS Number:
Molecular Weight:
646.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3[o+]c2-c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C27H30O16.ClH/c28-7-17-19(33)21(35)23(37)26(42-17)40-15-5-10(30)4-14-11(15)6-16(25(39-14)9-1-2-12(31)13(32)3-9)41-27-24(38)22(36)20(34)18(8-29)43-27;/h1-6,17-24,26-29,33-38H,7-8H2,(H2-,30,31,32);1H/t17-,18-,19-,20-,21+,22+,23-,24-,26-,27-;/m1./s1

InChI key

QDVZZZBBPRFPDG-DHJOXOLYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: primula sambucus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arshad Mehmood et al.
Combinatorial chemistry & high throughput screening, 23(9), 917-930 (2020-04-29)
The present study was designed to evaluate the xanthine oxidase (XO) inhibitory and antioxidant activities of 30 bioactive compounds present in edible food plants for the possible treatment of hyperuricemia. The XO inhibitory, SO and DPPH radical scavenging activities of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service