Skip to Content
Merck
All Photos(1)

Key Documents

BCR550

Glutaraldehyde 2,4-dinitrophenylhydrazone

ERM®, certified reference material

Synonym(s):

Glutaraldehyde bis(2,4-dinitrophenylhydrazone)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N8O8
CAS Number:
Molecular Weight:
460.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(N\N=C\CCC\C=N/Nc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O)c(c1)[N+]([O-])=O

InChI

1S/C17H16N8O8/c26-22(27)12-4-6-14(16(10-12)24(30)31)20-18-8-2-1-3-9-19-21-15-7-5-13(23(28)29)11-17(15)25(32)33/h4-11,20-21H,1-3H2/b18-8-,19-9+

InChI key

QAUJHIMXYLFORD-NCEWWTOFSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CRM47175CRM47345CRM47340
format

neat

format

single component solution

format

single component solution

format

single component solution

grade

certified reference material

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

manufacturer/tradename

JRC

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

agency

ERM®

agency

-

agency

-

agency

-

Analysis Note

For more information please see:
BCR550

Legal Information

ERM is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service