69888
Molybdate reagent solution
for photometric phosphate determination acc. to DIN 38405-D11-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
NA.21
Recommended Products
form
liquid
Quality Level
quality
for photometric phosphate determination acc. to DIN 38405-D11-1
technique(s)
UV/Vis spectroscopy: suitable
Related Categories
Components
solution contains hexaammonium heptamolybdate ((NH4)6Mo7O24.4H2O); sulfuric acid (H2SO4); potassium antimony(III) oxide tartrate(K(SbO)C4H4O6.0.5H2O);water
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hwan D Kim et al.
ACS applied materials & interfaces, 9(26), 21639-21650 (2017-06-14)
Chondroitin sulfate (CS) is the major component of glycosaminoglycan in connective tissue. In this study, we fabricated methacrylated PEGDA/CS-based hydrogels with varying CS concentration (0, 1, 5, and 10%) and investigated them as biomineralizing three-dimensional scaffolds for charged ion binding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service