35387
Ternidazole hydrochloride
VETRANAL®, analytical standard
Synonym(s):
1-(3-Hydroxypropyl)-2-methyl-5-nitro-1H-imidazole hydrochloride, 2-Methyl-5-nitroimidazole-1-propanol hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H11N3O3 · HCl
CAS Number:
Molecular Weight:
221.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
Cl.Cc1ncc(n1CCCO)[N+]([O-])=O
InChI
1S/C7H11N3O3.ClH/c1-6-8-5-7(10(12)13)9(6)3-2-4-11;/h5,11H,2-4H2,1H3;1H
InChI key
PTXLKVODAFRGDG-UHFFFAOYSA-N
Related Categories
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Recommended products
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Legal Information
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service