Skip to Content
Merck
All Photos(1)

Key Documents

236756

Supelco

Silica gel Inorganic Sorbent

Davisil grade 710, 1000-1400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Silica gel, pore size 50-76 Å, suitable for thin layer chromatography (TLC)

Assay

99%

Quality Level

form

powder

technique(s)

LPLC: suitable
thin layer chromatography (TLC): suitable

surface area

475-560 m2/g

matrix

Silica

matrix active group

silica

avg. part. size

10.0-14.0 μm

pore size

0.7-0.9 cm3/g pore volume
50-76 Å

pH

6.0-7.0 (5% in slurry)

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silica gel may be used as an adsorbent coated on a stationary phase used in the detection of acetylcholinesterase inhibitors from Amaryllidaceae extracts using thin layer chromatography (TLC).[1]

Other Notes

ph 6.0-7.0 5% slurry area 475-560 m2/g, 0.7-0.9 cc/g pore volume in addition to name changes

Legal Information

Davisil is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening for acetylcholinesterase inhibitors from Amaryllidaceae using silica gel thin-layer chromatography in combination with bioactivity staining
Rhee KI, et al.
Journal of Chromatography A, 915(1-2), 217-223 (2001)
T Martin et al.
Chemical communications (Cambridge, England), (1)(1), 24-25 (2002-07-18)
Texture-related features of water intrusion in hydrophobised MCM-41 silicas render these materials especially suitable for energy dissipation in mechanical dampers.
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Are Davisil series 710 silica gels heat activated?  What does "NW" in supplier name for product "Davisil Grade 710NW" refer to for product 236756?

    1 answer
    1. All of our Davisil silica gels are heated to dry just prior to packaging.  However, silica can easily pick up moisture when a container is opened.For any water sensitive applications, this is what we recommend:  Prior to use, to get rid of free water from the silica gel, dry the silica at 180°C for a few hours (e.g., 3 hours) in a drying oven. Note that at a temperature above 200°C, there will be loss of silanol groups.The "NW" is an internal Grace designation which is not necessarily related to the product other than it being a narrow particle size distribution and manufactured at a plant in Worms, Germany.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service