21496
Hamilton® syringe, 800 series, removable needle
805RN, volume 50 μL, needle size 22s ga (bevel tip), needle L 51 mm (2 in.)
All Photos(2)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
stainless steel needle (point style 2)
needle size
22s ga (bevel tip)
feature
removable needle RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 84857
needle L
51 mm (2 in.)
volume
50 μL
compatibility
for use with Hamilton 805RN (removable needle)
Related Categories
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)