Skip to Content
Merck
All Photos(1)

Key Documents

23-0310

Sigma-Aldrich

Coconut oil from Cocos nucifera

CP

Synonym(s):

Coconut fat, Copra oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211

grade

CP

form

solid

availability

available only in Japan

mp

23-27 °C

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aduja Naik et al.
Applied biochemistry and biotechnology, 167(5), 1290-1302 (2012-03-22)
Virgin coconut oil (VCO) has been gaining popularity in recent times. During its production, byproducts such as coconut skim milk and insoluble protein are obtained which are underutilized or thrown away to the environment at present. This study deals with
Quantitative measurement of the penetration of coconut oil into human hair using radiolabeled coconut oil.
V Gode et al.
Journal of cosmetic science, 63(1), 27-31 (2012-04-11)
Aida Serra et al.
Nutrition (Burbank, Los Angeles County, Calif.), 28(11-12), 1165-1171 (2012-08-04)
Phase II biotransformation of flavonoids generates bioactive metabolites in vivo. However, data on the effect of environmental and physiologic factors and fetal programming on phase II pathways toward flavonoids are limited. We examined the effect of parental exposure to a
A K Patra et al.
Journal of dairy science, 96(3), 1782-1792 (2013-01-22)
Coconut (CO) and fish (FO) oils were previously shown to inhibit rumen methanogenesis and biohydrogenation, which mitigates methane emission and helps improve beneficial fatty acids in meat and milk. This study aimed at investigating the comparative effects of CO and
ALSUntangled 15: coconut Oil.
Amyotrophic lateral sclerosis : official publication of the World Federation of Neurology Research Group on Motor Neuron Diseases, 13(3), 328-330 (2012-04-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service