Skip to Content
Merck
All Photos(1)

Key Documents

05-1530

Sigma-Aldrich

Carbon black

CP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

pellets
powder

availability

available only in Japan

particle size

24 nm (graphitized carbon black)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Actionable cancer vulnerability due to translational arrest, p53 aggregation and ribosome biogenesis stress evoked by the disulfiram metabolite CuET.
Kanellis, et al.
Cell Death and Differentiation, 30, 1666-1678 (2023)
Götz A Westphal et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 997-1005 (2015-04-22)
Inflammation is a decisive pathophysiologic mechanism of particle toxicity and accumulation of neutrophils in the lung is believed to be a crucial step in this process. This study describes an in vitro model for investigations of the chemotactic attraction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service