MIAC01401
Microfil Support with Valve 1/4in.
Clinical Lab, Food and Beverage, Industrial QC, Environmental Analysis, Laboratory Filtration
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.11
Recommended Products
Quality Level
manufacturer/tradename
Microfil® with S-Pak®
input
sample type beverage(s)
sample type food(s)
application(s)
bioburden testing
clinical
environmental
food and beverages
industrial qc
water monitoring
shipped in
ambient
Related Categories
General description
Microfil devices offer a convenient monitoring method to detect contamination related problems easily and reliably.
Legal Information
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service