MABT1353
Anti-C-Nap1 (Cep250) Antibody, clone 150-230-1
clone 150-230-1, from mouse
Synonym(s):
Centrosome-associated protein CEP250, 250 kDa centrosomal protein, Cep250, Centrosomal Nek2-associated protein 1, C-Nap1, Centrosomal protein 2
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
150-230-1, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
immunofluorescence: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... CEP250(11190)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected C-Nap1 (Cep250) in HeLa S3 cell lysates. (Courtesy of Prof. Erich A. Nigg, Ph.D., Biozentrum, University of Basel, Switzerland).
Quality
Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service