Skip to Content
Merck
All Photos(3)

Key Documents

MABS1182

Sigma-Aldrich

Anti-RagA/B Antibody, clone 1H7.1

clone 1H7.1, from mouse

Synonym(s):

Ras-related GTP-binding protein A, Adenovirus E3 14.7 kDa-interacting protein 1, FIP-1, Rag A, RagA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H7.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RRAGA(10670)

General description

Ras-related GTP-binding protein A (UniProt Q7L523; also known as Adenovirus E3 14.7 kDa-interacting protein 1, FIP-1. Rag A, RagA) is encoded by the RRAGA gene (Gene ID 10670) in human. Mammalian cells express four Rag GTPases that mediate amino acid sufficiency-dependent mTORC1 activation to support cell growth and proliferation. Rag GTPases assemble into heterodimers containing GTP-loaded RagA or RagB partnered with GDP-loaded RagC or RagD. Promoted by signal from an upstream amino acid-responsive supercomplex, the active Rag heterodimer accumulates on the cytoplasmic surface of the lysosome, where it recruits mTORC1 and initiates its activation. The pentameric Ragulator complex tethers Rag heterodimers to the lysosomal membrane and acts as RagA/B guanine nucleotide exchange factor (GEF), while the heterotrimeric GATOR1 complex and the heterodimeric FNIP-Folliculin complex function as RagA/B and RagC/D GTPase activating protein (GAP), respectively. Both lysosomal amino acid sensors, such as the multisubunit vacuolar ATPase (v-ATPase) and lysosomal amino acid transporter(s), and cytoplasmic amino acid sensors, such as the leucyl-tRNA synthetase (LeuRS), are part of the upstream amino acid-responsive supercomplex.

Specificity

Clone 1H7.1 targets an epitope within the C-terminal end sequence highly conserved between RagA and RagB.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminal end sequence of human/mouse/rat RagA.

Application

Detect RagA and RagB using this mouse monoclonal Anti-RagA/B, clone 1H7.1, Cat. No. MABS1182, validated for use in ELISA, Immunocytochemistry, and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected exogenously overexpressed human RagA in 1 µg of lysate from transfected HEK293T cells.

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected RagA/B immunoreactivity in HUVECs.

ELISA Analysis: Clone 1H7.1 hybridoma culture supernatant detected exogenously overexpressed human RagA and RagB in lysates (100 ng/well) from transfected HEK293T cells.

Quality

Evaluated by Western Blotting in HEK293.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected RagA in 10 µg of HEK293 cell lysate.

Target description

~36 kDa observed. 36.57 kDa (human/mouse/rat/bovine RagA), 43.25/40.17 kDa (human RagB Long/Short isoform), 43.19 kDa (mouse RagB), 43.19/40.11 (rat RagB Long/Short isoform) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond Liang et al.
Cell stem cell, 26(3), 359-376 (2020-02-29)
Quiescence is a fundamental property that maintains hematopoietic stem cell (HSC) potency throughout life. Quiescent HSCs are thought to rely on glycolysis for their energy, but the overall metabolic properties of HSCs remain elusive. Using combined approaches, including single-cell RNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service