Skip to Content
Merck
All Photos(1)

Key Documents

MAB5228

Sigma-Aldrich

Anti-Amyloid Precursor Protein Antibody, Secretory, clone OM84

clone OM84, Chemicon®, from mouse

Synonym(s):

APP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

OM84, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

Reacts with secretory amyloid precursor protein (sAPP), but not with Amyloid-Beta protein 1-40.

Immunogen

Amyloid Precursor Protein (APP) secreted by the EJ-1 bladder carcinoma cell line.
Epitope: Secretory

Application

Anti-Amyloid Precursor Protein Antibody, Secretory, clone OM84 detects level of Amyloid Precursor Protein & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blot: 10 μg/mL ELISA: 1 μg/mL

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 10 mM sodium phosphate buffer (pH 7.4), 0.15 M NaCl, and 0.1% sodium azide as a preservative..
Protein A purified

Storage and Stability

Maintain frozen at -70°C or below in undiluted aliquots for up to 12 months, avoid repeated freeze/thaw cycles.

Analysis Note

Control
Brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Römer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(20), 7777-7794 (2015-05-23)
Stroke-induced immunodepression (SIDS) is an essential cause of poststroke infections. Pharmacological inhibition of SIDS appears promising in preventing life-threatening infections in stroke patients. However, SIDS might represent an adaptive mechanism preventing autoreactive immune responses after stroke. To address this, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service