EZFITEXTE3
EZ-Fit® Manifold Filtration Head Extension
for use with EZ-Fit® Filtration Manifold
Synonym(s):
EZ-Fit® Filtration Manifold Accessory, EZ-Fit® Manifold
About This Item
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
packaging
pkg of 3 units
manufacturer/tradename
EZ-Fit®
input
sample type bottled water
sample type cosmetics
sample type pharmaceutical(s)
sample type beer
application(s)
bioburden testing
cosmetics
food and beverages
pharmaceutical
water monitoring
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
configured for
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service