B485-1L
Rinse Fluid A
bottle of 1 L, pkg of 4 units, square polycarbonate bottle (1L with Standard Cap), sterile; autoclaved
Synonym(s):
Fluid A for Sterility Testing, Rinse Fluid, Rinsing Solution Fluid A, Washing Fluid
About This Item
Recommended Products
material
square polycarbonate bottle (1L with Standard Cap)
Quality Level
Agency
EP 2.6.13
JP 4.06
USP 71
sterility
sterile; autoclaved
form
liquid
packaging
bottle of 1 L
pkg of 4 units
technique(s)
microbiological culture: suitable
sterility testing: suitable
pH range
6.9-7.3
application(s)
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for bacteria
(
)
Looking for similar products? Visit Product Comparison Guide
General description
Fluid A washing solution in general serves as a rinse buffer and can be used for an initial rinse of the filter membrane to inactivate any inhibitors, if present.
Application
Analysis Note
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service