Skip to Content
Merck
All Photos(1)

Key Documents

AB26

Sigma-Aldrich

Goat Anti-Mouse IgG (10mL) KC Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

serum

Quality Level

clone

polyclonal

species reactivity

mouse, mouse, mouse

technique(s)

immunoprecipitation (IP): suitable

Storage and Stability

Store frozen at -20° C in undiluted aliquots for up to 12 months.Avoid multiple freeze-thaws.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service