Skip to Content
Merck
All Photos(1)

Key Documents

8.43793

Sigma-Aldrich

Propionic acid sodium salt

for synthesis

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NaO2
CAS Number:
Molecular Weight:
96.06
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-290-4
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

solid

potency

5600 mg/kg LD50, oral (Rat)
1640 mg/kg LD50, skin (Rabbit)

pH

8-9.5 (20 °C, 10 g/L in H2O)

mp

289.3 °C

solubility

>500 g/L

density

1.5103 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service