345797
α1-3,6-Galactosidase, Recombinant, E. coli
Catalyzes the hydrolysis of α1,3-linked and α1,6-linked, non-reducing terminal galactose from complex carbohydrates and glycoproteins.
Synonym(s):
α1-3,6-Galactosidase, Recombinant, E. coli, α1-3,6-D-Galactoside Galactohydrolase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Recommended Products
recombinant
expressed in E. coli
Quality Level
form
liquid
specific activity
≥30 units/mg protein
≥85 units/mL
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
foreign activity
Proteases, none detected
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
Recombinant α1-3,6-galactosidase expressed in E. coli. Catalyzes the hydrolysis of α1,3-linked and α1,6-linked, non-reducing terminal galactose from complex carbohydrates and glycoproteins. Particularly efficient in removing α-linked galactose under conditions where the pH must be neutral or above, as with live cells.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will catalyze the release of 1.0 µmol p-nitrophenol from p-nitrophenyl-α-D-galactopyranoside per min at 25°C, pH 6.5.
Physical form
In 50 mM sodium phosphate buffer, pH 7.5.
Other Notes
Prime, S., et al. 1996. J. Chromatogr. 720, 263.
Dwek, R.A., et al. 1993. Annu. Rev. Biochem. 62, 65.
Dwek, R.A., et al. 1993. Annu. Rev. Biochem. 62, 65.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service