Skip to Content
Merck
All Photos(1)

Key Documents

1.93102

Supelco

Sodium hydroxide

pellets (low chloride), for analysis EMPARTA® ACS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352316

Product Name

Sodium hydroxide pellets (low chloride), for analysis EMPARTA® ACS

grade

ACS reagent

product line

EMPARTA®

Assay

99%

form

solid

pH

> 14 (20 °C, 100 g/L)

bp

1,390 °C/1,013 hPa

mp

318  °C

solubility

water: soluble 1,090 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

anion traces

chloride (Cl-): 0.005%
phosphate (PO43-): 0.001%
sulfate (SO42-): 0.003%

cation traces

Ca: 0.005%
Fe: 0.001%
K: 0.05%
Mg: 0.002%
N: 0.001%
Ni: 0.001%
heavy metals (as Pb): 0.002%

storage temp.

20-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Packaging

1.93102.0521: 500G, Minimum order quantity: 20

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service