Skip to Content
Merck
All Photos(1)

Key Documents

16-293

Sigma-Aldrich

Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Alexa Fluor 488

clone SKB1, Upstate®, from mouse

Synonym(s):

PKB, PKD-alpha, AKT1, Protein Kinase B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

SKB1, monoclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Akt1(11651)

Related Categories

Specificity

Recognizes Akt1/PKBα > Akt2/PKBβ (30%) > Akt3/PKBγ (weak to none) based on immunoblot analysis using 100ng recombinant fusion protein for each isoform.

Immunogen

GST-fusion protein corresponding to residues 1-149 of human Akt1. Clone SKB1.

Application

Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Alexa Fluor 488 is an antibody against Akt/PKB for use in FC, IC, IF & WB.

Quality

Routinely evaluated by Immunocytochemistry and Flow Cytometry

Target description

60 kDa

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elke Butt et al.
Cells, 9(2) (2020-02-23)
The serine/threonine protein kinase AKT1 is a downstream target of the chemokine receptor 4 (CXCR4), and both proteins play a central role in the modulation of diverse cellular processes, including proliferation and cell survival. While in chronic myeloid leukemia (CML)
Chanya Inprasit et al.
International journal of molecular medicine, 45(3), 779-792 (2020-01-11)
Obesity is a global health problem affecting the general population. Acupoint catgut embedding (ACE) is an alternative treatment that involves the implantation of absorbable catgut suture at acupoints. The transient receptor vanilloid member 1 (TRPV1) is a calcium ion channel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service