Skip to Content
Merck
All Photos(1)

Key Documents

1.15934

Sigma-Aldrich

Eosin B (bluish) (C.I.45400)

for microscopy Certistain®

Synonym(s):

Eosin B (bluish) (C.I.45400), Dibromodinitrofluorescein sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br2N2Na2O9
CAS Number:
Molecular Weight:
624.06
MDL number:
UNSPSC Code:
41116130
EC Index Number:
208-943-1

Quality Level

form

solid

composition

Dye, ≥85% spectrophotometric assay

pH

6.2 (25 °C, 10 g/L in H2O)

solubility

300 g/L

bulk density

680 kg/m3

εmax

800-1000 at 517-525 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H8Br2N2O9.2Na/c21-14-16(25)11(23(29)30)5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(24(31)32)17(26)15(22)19(10)33-18(9)14;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

GYYTYUGGVBYJHE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Eosin B (C.I. 45400) for microscopy Certistain®, is a dry dye for microscopy that is often used as a counterstain, vizualising proteins, connective tissues, fibers and keratin in sample material of human origin. Depending on user requirements and personal preferences, it can be used for the preparation of the commonly used aqueous or alcoholic Eosin B staining solutions. This staining solution will mostly be used for routine staining in the hematoxylin-eosin staining process (H&E). The H&E staining process is the most frequently used staining method for histological material. Due to the hematoxylin solution, the nuclei will appear dark blue to dark violet in color and the Eosin Y solution will stain the cytoplasma and intercellular substances pink to red. As the dye belongs to the portfolio of Certistain® dyes, it is chemically analyzed according to strict specifications before release (CE marked and IVD product) and a high staining quality, with reliable staining results is guaranteed. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (gravimetrically): ≥ 85 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 517 - 525 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 800 - 1000
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service