1.14413
Aluminum Test Kit, colorimetric
0.07-0.80 mg/L (Al), MQuant®
Synonym(s):
Aluminium Visual Test Kit
About This Item
Recommended Products
Product Name
Aluminium Test, colorimetric, 0.07-0.80 mg/L (Al), for use with MQUANT®
specific analyte(s)
aluminium
Quality Level
measuring range
0.07-0.80 mg/L (Al)
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
Method
In weakly acidic, acetate-buffered solution aluminium ions react with chromazurol S to form a blue compound. Due to the intrinsic orange coloration of the reagent blank, the measurement solution is orange-red to red in color. The aluminium concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.18452 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical system.
This test can be used in
- Groundwater and surface water, seawater
- Drinking water and mineral water
- Waters from aquaculture
- Boiler and boiler feed water, cooling water
- Industrial and process water
- Wastewater and percolating water
- Electroplating wastewater
- Swimming-pool water
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service