Skip to Content
Merck
All Photos(1)

Key Documents

1.01722

Supelco

Barium fluoride

99.99 Suprapur®

Synonym(s):

Barium fluoride, Barium difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaF2
CAS Number:
Molecular Weight:
175.32
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-108-0
Assay:
99.99%
grade:
for inorganic trace analysis
form:
solid
solubility:
1.2 g/L

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

250 mg/kg LD50, oral (Rat)

bp

2260 °C/1013 hPa

mp

1353 °C

solubility

1.2 g/L

density

4.89 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/Ba.2FH/h;2*1H/q+2;;/p-2

InChI key

OYLGJCQECKOTOL-UHFFFAOYSA-L

Related Categories

Analysis Note

Purity (metallic): ≥ 99.99 %
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 0.5 ppm
Co (Cobalt): ≤ 0.5 ppm
Cu (Copper): ≤ 0.5 ppm
Fe (Iron): ≤ 0.5 ppm
K (Potassium): ≤ 20 ppm
Li (Lithium): ≤ 5 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 0.5 ppm
Na (Sodium): ≤ 50 ppm
Ni (Nickel): ≤ 0.5 ppm
Pb (Lead): ≤ 0.5 ppm
Tl (Thallium): ≤ 0.5 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service