Skip to Content
Merck
All Photos(1)

Key Documents

H-142

Supelco

4-Hydroxyalprazolam solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4O
CAS Number:
Molecular Weight:
324.76
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C17H13ClN4O/c1-10-20-21-16-17(23)19-15(11-5-3-2-4-6-11)13-9-12(18)7-8-14(13)22(10)16/h2-9,17,23H,1H3

InChI key

CEYGCFLPQFNPST-UHFFFAOYSA-N

General description

An analytical reference standard suitable for use as calibrators or controls in a variety of LC/MS or GC/MS applications such as clinical toxicology, or urine drug testing. 4-Hydroxyalprazolam is a major metabolite of alprazolam, detected in serum or in urine. 4-hydroxyalprazolam can be further metabolized to benzophenone.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.0 °F - closed cup

Flash Point(C)

12.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service