Skip to Content
Merck
All Photos(1)

Key Documents

Z742922

Tergajet® detergent, low foaming phosphate-free powder

pkg of 1.8 kg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pkg of 1.8 kg

manufacturer/tradename

Alconox, Inc. 2204

application(s)

detection

Looking for similar products? Visit Product Comparison Guide

General description

Tergajet low-foaming powdered detergent

  • Concentrated to save you money
  • Phosphate-free, biodegradable and readily disposable
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Low foaming heavy duty detergent for high performance results
  • Free rinsing to give you reliable results and no interfering residues
  • Ideal for under-counter and floor standing lab washing machines

Used to clean: Water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning
compliant. FDA certified.

Used to remove: Soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.

Surfaces cleaned: Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for Aluminum. Corrosion testing is advisable.

Cleaning method: Laboratory warewashers, dishwashers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-inplace and manual cleaning if proper skin and eye protection is used.

Directions: Follow machine manufacturer′s directions and use the powder detergent receptacles. In general, use 1/2 - 1% hot water wash cycle solutions (1 1/4 - 2 1/2 Tbsp. per gal., 3/4 - 1 1/4 oz. per gal. or 5-10 grams per liter. Most machines need 2 1/2 oz. added at a typical 2 gal. wash cycle).
For manual, soak, circulate or ultrasonic use, make a 1% solution in warm or hot water.
For abrasive scrubbing, make a paste.
For difficult soils, raise water temperature and use more detergent. Wear protective gloves and eyewear when handling.
RINSE THOROUGHLY--preferably with running water. For critical cleaning, do final or all rinsing in distilled, deionized or purified water. For food contact surfaces, rinse with potable water.

Legal Information

Tergajet is a registered trademark of Alconox, Inc.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service