Z681768
MyFuge™ 12 Mini Centrifuge Adapters
to hold, 0.2 mL tubes
About This Item
Recommended Products
feature
to hold 0.2 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Benchmark Scientific C1008-A2-6
Benchmark Scientific
technique(s)
centrifugation: suitable
compatibility
for use with MC-12 High Speed Microcentrifuge
for use with MC-24 High Speed Microcentrifuge
for use with myFuge 12 Mini Centrifuge
for use with myFuge 5 MicroCentrifuge
for use with myFuge Mini Centrifuge
Related Categories
General description
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)