Key Documents
Z680850
Duran® stirred reactor caps and accessories
spare screw cap, 2-ports
Synonym(s):
1200390
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
DURAN®
Schott
fitting
for for GL 45 stirred reactor systems
General description
These components allow for extra media bottles to be connected or lets sterile pressure compensation be possible. The parts are autoclaveable and the drive is provided by a standard magnetic stirrer.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)