Z408727
Ace gas dispersion tube with porous fritted glass tip
Fit #7 Ace-Thred, O.D. × L 7 mm × 135 mm, porosity 10-20 μm (D)
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
manufacturer/tradename
Ace Glass 943524
O.D. × L
7 mm × 135 mm
pore size
10-20 μm porosity (D)
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)