Z292044
Open-top style aluminum caps
open center, 9.5 mm opening, For autosampler headspace analysis, pkg of 1000 ea
Synonym(s):
Aluminum crimp seal (without septum), silver aluminum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
silver aluminum
feature
open center 9.5 mm opening
packaging
pkg of 1000 ea
diam.
20 mm
compatibility
for use with 20 mm crimp seal vial
Looking for similar products? Visit Product Comparison Guide
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service