Z291811
WISP™ 48-position autosampler vials
clear screw-thread vials, pkg of 100 ea, thread for 13-425
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear borosilicate glass vial
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
15 mm × 45 mm
volume
4 mL
4 mL
fitting
thread for 13-425
Looking for similar products? Visit Product Comparison Guide
General description
Constructed of borosilicate glass. Screw-thread vials use 11 mm septa and 13-425 threaded screw-caps.
Suitability
Autosampler compatibility:
Agilent 7673A, Series I, II
Shimadzu LC Autosamplers
Waters 48, 717, 717 Plus
Agilent 7673A, Series I, II
Shimadzu LC Autosamplers
Waters 48, 717, 717 Plus
Legal Information
WISP is a trademark of Waters
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service