Skip to Content
Merck
All Photos(1)

Key Documents

Z267686

Sigma-Aldrich

Round crystal window

KRS-5, diam. × thickness 25 mm × 2 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

material

KRS-5

technique(s)

IR spectroscopy: suitable

diam. × thickness

25 mm × 2 mm

transmittance

20,000-25,000 (wavenumbers (cm-1))

refractive index

n/D 2.37 (at 2 μm)

Looking for similar products? Visit Product Comparison Guide

General description

A soft crystal, deforms under pressure; good ATR material.
Cut and polished quality crystal windows.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service