Z189006
Ace four-neck round-bottom flask
capacity 500 mL
Synonym(s):
bottom flask, flask round, flask round bottom
About This Item
Recommended Products
material
borosilicate glass
four-neck
round bottom flask
Joint
90 degree angle joint ST/NS24/40
center joint ST/NS34/45 vertical
side Joints ST/NS24/40 vertical, except for 500 mL, which is angled (2)
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6952-04
capacity
500 mL
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)