Skip to Content
Merck
All Photos(1)

Key Documents

W203807

Sigma-Aldrich

Allyl phenoxyacetate

≥99%, FG

Synonym(s):

2-propenyl phenoxyacetate, Allyl 2-phenoxyacetate, prop-2-enyl 2-(phenoxy)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
192.21
FEMA Number:
2038
EC Number:
Council of Europe no.:
228
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.701
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.516 (lit.)

bp

265-266 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

honey; pineapple; sweet

SMILES string

C=CCOC(=O)COc1ccccc1

InChI

1S/C11H12O3/c1-2-8-13-11(12)9-14-10-6-4-3-5-7-10/h2-7H,1,8-9H2

InChI key

VUFZVGQUAVDKMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • In situ synthesis of a monolithic material with multi-sized pores and its chromatographic properties for the separation of intact proteins from human plasma.: The study discusses the development of a new chromatographic material that improves protein separation, which is crucial for biochemical analysis and diagnostics (Lan et al., 2019).

  • The effects of vehicles on the human dermal irritation potentials of allyl esters.: This investigation focuses on the dermal irritation potential of allyl esters, including allyl phenoxyacetate, and the impact of different vehicles on irritation levels, relevant for developing safer personal care products (Politano et al., 2006).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facial dermatitis from allyl phenoxyacetate in a dry carpet shampoo.
A E Taylor et al.
Contact dermatitis, 8(1), 70-70 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service