Key Documents
SYNP120010M
Synthware peptide synthesis vessel with fritted disc and PTFE stopcock
10 mL, disc O.D. 15 mm, porosity: medium, GL 14
Synonym(s):
Synthware P120010M, Solid phase peptide synthesis vessel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P120010M
capacity
10 mL
10 mL
disc O.D.
15 mm
fitting
GL 14
Related Categories
General description
With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)