Key Documents
SYNF318250
Synthware recovery flask
250 mL, joint: ST/NS 29/42
Synonym(s):
Synthware F318250, oval shape flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F318250
capacity
250 mL
250 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)