Skip to Content
Merck
All Photos(1)

Key Documents

JKA4077

Sigma-Aldrich

Vinylsulfone-PEG-Vinylsulfone

average MN 5,000, cross-linking reagent thiol reactive

Synonym(s):

Vinylsulfone-PEG-Vinylsulfone, VS-PEG-VS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CHSOOCH2CH2O(CH2CH2O)nCH2CH2SOOCHCH2
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Vinylsulfone-PEG5K-Vinylsulfone, average Mn 5,000

form

solid

mol wt

average Mn 5,000

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

General description

Vinylsulfone PEG Vinylsulfone. Sulfhydryl reactive PEG crosslinker.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service