DWKBGE342P
Azlon Square Bottles
graduated, cap, Plug, translucent high-density polyethylene, capacity 2000 mL
Synonym(s):
BGE342P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121818
NACRES:
NB.43
material
translucent high-density polyethylene
feature
cap Plug
graduated
packaging
pkg of 5 ea
manufacturer/tradename
Azlon
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for storage of liquid and powder samples
Features and Benefits
Space saving square design
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service