Skip to Content
Merck
All Photos(1)

Key Documents

CLS894719

Pyrex® vacuum connecting adapter, 105° angle

bottom male joint: ST/NS 19/22, top female joint: ST/NS 19/22

Synonym(s):

Corning® 8947

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.43

Joint

female joint: ST/NS top19/22
male joint: ST/NS bottom19/22

manufacturer/tradename

Corning 8947-19

Related Categories

General description

With tubulation of 10mm O.D. for attaching to a vacuum pump or to inner and outer joints. Includes a drip-tip on the inner tube.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service