CLS71036
Pyrex® volumetric pipette, to contain, to deliver, Class A
orange, volume 6 mL, accuracy: 0.01 mL
Synonym(s):
Corning® 7103
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass
feature
class A
graduation
packaging
case of 6 ea
pack of 6 ea
manufacturer/tradename
Corning 7103-6
volume measuring range
6 mL, accuracy: 0.01 mL
L
420 mm
capacity
6 mL
color
orange
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® volumetric pipettes offer high precision measurement of liquids.
- Calibrated "To Contain" (bottom line) and "To Deliver" (top line) to specifications for Class A.
- Color-coded with colored graduations for easy identification.
- Calibration lines enameled onto the glass to ensure volume accuracy.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service