Key Documents
CLS70980500
Pyrex® VISTA Erlenmeyer flask, narrow-mouth
capacity 500 mL
Synonym(s):
conical flask, conical flasks, erlenmeyer flasks, flask erlenmeyer, Corning® 70980
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
narrow-mouth
sterility
non-sterile
feature
cap: no
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70980-500
capacity
500 mL
General description
These narrow mouth Erlenmeyer flasks have uniform wall thickness which provide the proper balance between mechanical strength and thermal shock resistance. Approximate graduations are in durable blue enamel. An extra large marking space is also provided.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)