CLS70640100
Pyrex® VISTA™ volumetric flask, Class A
capacity 100 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 70640
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
class A
closure type ST stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70640-100
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Pyrex® VISTA™ volumetric flasks, Class A (volume 100 mL) provide precise volume measurement.
- These flasks have sealed necks tooled for No. 13 Standard Taper glass stoppers.
- The graduation line is sharp and permanent and large blue block letters make the inscription easy to read.
- These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks.
- This product will withstand repeated standard autoclave cycles of 121°C at 15 psi for 15 min.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service