Skip to Content
Merck
All Photos(1)

Key Documents

CLS69444L

Pyrex® rectangular chromatography jar

L × W × H 137 mm × 162 mm × 267 mm

Synonym(s):

Corning® 6944

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

grade

for chromatography

manufacturer/tradename

Corning 6944-4L

L × W × H

137 mm × 162 mm × 267 mm

capacity

3.8 L

Looking for similar products? Visit Product Comparison Guide

General description

Ground to the close tolerances needed for tight cover fit. Jar edges are ground flat within 0.25mm. Covers are not supplied. Do not use heat, pressure or vacuum applications.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service