Skip to Content
Merck
All Photos(1)

Key Documents

CLS5640200FO

Pyrex® class A volumetric flask with ST joint, flask only

capacity 200 mL

Synonym(s):

flasks glass, volumetric flasks, volumetric glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804

feature

class A

packaging

case of 1 ea

manufacturer/tradename

Corning 5640-200FO

tol. measuring range

0.10 mL

capacity

200 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Pyrex® class A volumetric flasks with ST joint, flask only (volume 200 mL) provide improved lab convenience, broad selection and precise volume measurement.
  • The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for No. 13 Standard Taper glass stoppers.
  • Stoppers are supplied separately.
  • The graduation line is sharp and permanent and large white block letters make the inscription easy to read.
  • These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service