Key Documents
CLS55020250
Pyrex® low actinic narrow mouth Erlenmeyer flask, Pyrex® ST 27 stopper
capacity 250 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flask with stopper, flasks erlenmeyer, Corning® 55020
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
narrow-mouth
sterility
non-sterile
feature
closure type ST stopper
packaging
case of 1
manufacturer/tradename
Corning 55020-250
capacity
250 mL
General description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of the flask, which retains the mechanical strength, chemical stability and thermal resistance of Pyrex brand labware.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)