Skip to Content
Merck
All Photos(1)

Key Documents

CLS33950MEC

Corning® PYREX® filter discs 1722 μm pore size

body diam. 35 mm, porosity 170-220 μm

Synonym(s):

Corning® 33950

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.43

product name

Pyrex® fritted disc, body diam. 35 mm, porosity 170-220 μm

manufacturer/tradename

Corning 33950-MEC

body diam.

35 mm

pore size

170-220 μm porosity

Looking for similar products? Visit Product Comparison Guide

General description

Designed for use in Pyrex brand extractors, where it is desirable to weigh the material extracted or where paper thimbles may be attacked by the chemicals used.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service