BMSR20201520
Benchmark RotoMini Rotator Tube holder
to hold, 11 x 1.5/2.0
Synonym(s):
tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
to hold 11 x 1.5/2.0
manufacturer/tradename
Benchmark Scientific R2020-1520
Looking for similar products? Visit Product Comparison Guide
Application
Mixing of samples in tubes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service