Key Documents
BMSH1000MR25
Benchmark MAGic ”Clamp” magnetic clamp
to hold, 25 mL Erlenmeyer
Synonym(s):
magnetic clip
About This Item
UNSPSC Code:
41122413
NACRES:
NB.13
feature
to hold 25 mL Erlenmeyer
manufacturer/tradename
Benchmark Scientific H1000-MR-25
compatibility
MAGic Clamp universal platform
General description
Unique magnetic clamps for use with the MAGic Clamp platforms for a convenient method attaching flasks or racks to a shaker.
Choose from one of the most recent versions:
Certificates of Analysis (COA)