Key Documents
BAF199170250
SP Bel-Art® HiFlow Vacuum Aspirator Collection System
AC/DC input 100 V / 220 V (Includes US and EU line cords)
Synonym(s):
HIFLOW VACUUM ASPIRATOR COLLECTION SYSTEM, HiFlow Pump System, SELF-CONTAINED HI-FLOW VACUUM ASPIRATOR SYSTEM, SP BEL-ART HIFLOW VACUUM ASPIRATOR COLLECTION SYSTEM, 1.0 GALLON BOTTLE WITH PUMP
About This Item
UNSPSC Code:
41121820
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F19917-0250
L × W × H
23.2 cm × 17.1 cm × 43.8 cm, 9 1/8 x 6¾ x 17¼“H
Related Categories
General description
This compact, portable system is ideal for vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes and is available with the original strength or HiFlow pump.
Features and Benefits
  • Self-contained system with 1.0 gallon bottle, tubing and vacuum pump is lightweight, compact and easy to use
  • Practical whenever a house vacuum is absent or not working
  • Built-in, maintenance free pump quickly and conveniently aspirates fluids
  • HiFlow model has a stronger flow rate to work reliably with up to 8-channel pipette tips, even with 4 channels empty
  • HiFlow model includes quick disconnects to allow easy removal, emptying and replacement of the bottle
  • Low noise level (76dbs or less at 1 meter); minimal pump vibration
  • Chemical and corrosion resistant polypropylene collection bottle features heavy wall construction and a safety release vent to prevent vacuum implosion
  • Replaceable hydrophobic filter protects the pump motor from fluid contamination
  • Collection bottle, lid, tubing and connectors are all autoclavable at 121°C; all held within a polypropylene pump housing
  • CE marked and CSA certified and the transformer includes an additional cord for 220V type C outlets
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)