BAF131920102
Clavies® cover for biohazard bag holder
pack of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
material
plastic
feature
Medium (F13192-0002) or Large (F13192-0003) Clavies Biohazard Bag Holders
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13192-0102
size
203x241 mm
Related Categories
General description
Dishwasher and autoclave safe, the reusable Clavies® Biohazard Bag Holder Cover opens quickly and easily by pressing the built-in release tab – no batteries required! The cover fits both the Medium and Large sizes of the Clavies® Biohazard Bag Holders and features a smooth 10.1 x 10.1cm (4 x 4 in.) area for applying labels.
- Opens with a single touch; sanitary, reusable
- Can be installed in seconds
- Improves the sanitation of these biohazardous waste containers
- Center panel of the lid opens to an upright position and reveals a large 20.3 x 24.1cm (8 x 9 1/2 in.) opening through which wastes can be deposited
- Autoclavable at 121°C (250°F)
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)